Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EOGT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553990 Supplier Invitrogen™ Supplier No. PA553990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEAPPNTHSI PGEPLYNYAS IRLPEEHIPF FLHNNRHIAT VCRKDSLCPY KKHLEKLKYC WGYEKSCKPE FRFGYPVCSY VDMGWTDTLE SAEDIFWKQA DFGYARERLE EMHVLCQPKE TSDSSLVCSR YLQ Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

The exact function of C3orf64 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EOGT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eogt
Gene Accession No. Q5NDL2
Gene Alias A130022J15Rik; Aer61; AER61 glycosyltransferase; AI447490; AOS4; AW214473; AW259391; C3orf64; EGF domain specific O-linked N-acetylglucosamine transferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF domain-specific O-linked N-acetylglucosamine transferase; EGF-O-GlcNAc transferase; EOGT; EOGT1; Extracellular O-linked N-acetylglucosamine transferase; glycosyltransferase Aer61
Gene Symbols Eogt
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EOGT. Recombinant protein control fragment (Product #RP-90815).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285203
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.