Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EOGT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117936
This item is not returnable.
View return policy
Description
EOGT Polyclonal Antibody for Western Blot, IP
The exact function of C3orf64 remains unknown.
Specifications
| EOGT | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q5NDL2 | |
| Eogt | |
| E. coli-derived Human AER61/C3orf64 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Eogt | |
| A130022J15Rik; Aer61; AER61 glycosyltransferase; AI447490; AOS4; AW214473; AW259391; C3orf64; EGF domain specific O-linked N-acetylglucosamine transferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF domain-specific O-linked N-acetylglucosamine transferase; EGF-O-GlcNAc transferase; EOGT; EOGT1; Extracellular O-linked N-acetylglucosamine transferase; glycosyltransferase Aer61 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 285203 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction