Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EOMES Monoclonal Antibody (WD1928), PE-Cyanine5.5, eBioscience™, Invitrogen™

Catalog No. 35487741
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-487-741 25 Tests
35-487-742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-487-741 Supplier Invitrogen™ Supplier No. 35487741
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EOMES
Applications Flow Cytometry
Classification Monoclonal
Clone WD1928
Concentration 5 μL/Test
Conjugate PE-Cyanine5.5
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Eomes
Gene Accession No. O95936
Gene Alias C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2
Gene Symbols Eomes
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100622125, 8320
Target Species Human, Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.