Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EOMES Mouse anti-Human, Mouse, PE, Clone: X4-83, BD Biosciences™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 566749

Catalog No. BDB566749


Only null left
Add to Cart

Description

Description

EOMES Monoclonal antibody specifically detects EOMES in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EOMES
Monoclonal
0.2 mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
0.1 mg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
Flow Cytometry
X4-83
PE
C77258; T-box brain protein 2; TBR-2; TBR2; Tbr2; eomesodermin homolog
Human EOMES aa 1-275 Recombinant Protein
RUO
Human, Mouse
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.