Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EOMES Recombinant Rabbit Monoclonal Antibody (BLR104H)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | EOMES |
---|---|
Clone | BLR104H |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544399
|
Invitrogen™
MA544399 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544400
|
Invitrogen™
MA544400 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.Specifications
EOMES | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
O95936 | |
8320 | |
Peptide representing a region between residue 125 and 175 of Human eomesodermin homolog (EOMES). | |
Antibody |
BLR104H | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
Eomes | |
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
Eomes | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title