Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium hydroxide, 25% in water

Catalog No. AC420295000
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium hydroxide
Color Colorless to Yellow
Name Note 25% in Water
CAS 77-98-5, 7732-18-5
Boiling Point 102.0°C
Molecular Formula C8H21NO
Linear Formula (C2H5)4NOH
MDL Number MFCD00009024
Density 1.0200g/mL
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
Solubility Information Solubility in water: soluble.
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES [OH-].CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;hydroxide
Molecular Weight (g/mol) 147.26
PubChem CID 6509
Formula Weight 147.26
Percent Purity 24 to 26%
Packaging Plastic bottle
Refractive Index 1.402
Specific Gravity 1.02
Quantity 500 g
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.