Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y, 1% Alcoholic, LabChem™

Catalog No. p-5389146
Click to view available options
Quantity:
500 mL
1 L

Specifications

Physical Form Liquid
Chemical Name or Material Eosin Y
Name Note 1% Alcoholic
CAS Max % 89.5, 5.25, 4.25, 1
CAS 67-63-0, 67-56-1, 17372-87-1
Packaging Poly Bottle
Solubility Soluble in water
Decomposition Information Carbon monoxide; Carbon dioxide; Hydrogen bromide
Color Green
Quantity 500 mL
Linear Formula C20H6Br4O5·2Na
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Formula Weight 691.86
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.