Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18602416 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-416 25 g Amber Glass Bottle
18-602-415 5 g Amber Glass Bottle
18-602-418 100 g Amber Glass Bottle
18-602-417 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-416 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E129525GM
Add to Cart
Add to Cart

E1295, 17372-87-1, C20H6Br4Na2O5

Spectrum™ Chemical Eosin Y, Reagent, ACS , a form of eosin, is used as an acidic red stain for indicating cytoplasm material. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H6Br4Na2O5
Molecular Weight (g/mol) 691.86
InChI Key SEACYXSIPDVVMV-UHFFFAOYSA-L
IUPAC Name disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12

Specifications

Physical Form Crystalline Powder
CAS Max % 100%
Packaging Amber Glass Bottle
Color Red to Brown
Quantity 25 g
Grade ACS
Chemical Name or Material Eosin Y
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.