Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y TS, 0.5% (w/v), Adsorption Indicator for Argentometric Titrations, Ricca Chemical
SDP

Catalog No. p-7239379 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Specifications

Chemical Name or Material Eosin Y
Name Note Test Solution, Adsorption Indicator For Argentometric Titrations
CAS 17372-87-1, 7732-18-5
Concentration 0.5% (w/v)
Molecular Formula C20H12Br4Na2O8
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
Molecular Weight (g/mol) Mixture
PubChem CID 91886399
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Dark Red
Quantity 120 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.