Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Eosinophil Cationic Protein, Mouse, Unlabeled, Clone: 1C8.5/D8,

Catalog No. MABF986MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF986MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF986MI Supplier MilliporeSigma™ Supplier No. MABF986
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Eosinophil cationic protein (UniProt P12724; also known as ECP, Ribonuclease 3, RNase 3) is encoded by the RNASE3 (also known as ECP, RNS3) gene (Gene ID 6037) in human. The major basic protein (MBP), eosinophil cationic protein (ECP), eosinophil derived neurotoxin (EDN) and eosinophil peroxidase (EPO), are eosinophil effector molecules implicated in host defense, immunoregulatory responses, and in allergic and inflammatory reactions. These highly cationic proteins are stored preformed within the eosinophil secondary granules and released in intact membrane-bound granules during cell lysis or through a complex system of vesiculotubular structures during “piecemeal necrosis”. While eosinophil count and eosinophil surface expression of the activation marker CD69 are significantly correlated with serum concentrations of MBP, EDN and EPO, studies show that they are not correlated with ECP level. On the other hand, eosinophilic disease patients with normal eosinophil count show significantly increased concentrations of MBP, ECP, EDN, and EPO compared to normal donors. The measurement of MBP, ECP, EDN, and EPO levels thus provides a better predictor than eosinophil count alone in disease diagnosis. ECP is produced with a signal peptide sequence (a.a. 1-27), the removal of which yields the mature (a.a. 28-160) protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eosinophil Cationic Protein
Applications Immunohistochemistry (Paraffin), Immunoassay, Western Blot
Classification Monoclonal
Clone 1C8.5/D8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene RNASE3, ECP, RNS3
Gene Accession No. P12724
Host Species Mouse
Immunogen Purified human ECP.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002926
Test Specificity Clone 1C8.5/D9 specifically reacted with ECP, but not other granule proteins by Western blotting analysis (Makiya, M.A., et al. (2014). J. Immunol. Methods. 411:11-22).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.