Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eosinophil derived neurotoxin Antibody (HL2166) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325453
Description
Eosinophil derived neurotoxin Monoclonal antibody specifically detects Eosinophil derived neurotoxin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Eosinophil derived neurotoxin | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HL2166 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.1.27.5, EDNRNase 2, Eosinophil-derived neurotoxin, non-secretory ribonuclease, Ribonuclease 2, Ribonuclease US, ribonuclease, RNase A family, 2 (liver, eosinophil-derived neurotoxin), RNS2RNase UpI-2 | |
Recombinant fragmemt of human Eosinophil derived neurotoxin. | |
100 μL | |
Protein Phosphatase | |
6036 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction