Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Eotaxin 2 Monoclonal Antibody (02), Invitrogen™

Catalog No. PIMA543504
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543504 50 μL
PIMA543009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543504 Supplier Invitrogen™ Supplier No. MA543504
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human Eotaxin-2/CCL24

Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eotaxin 2
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL24
Gene Accession No. O00175
Gene Alias CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24
Gene Symbols CCL24
Host Species Mouse
Immunogen Recombinant Human Eotaxin-2/CCL24 Protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6369
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.