Learn More
Invitrogen™ Eotaxin 2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Supplier: Invitrogen™ 500P103GBT500UG

Description
AA Sequence of recombinant protein: VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVA. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin-2 (CCL24). Anti-Human Eotaxin-2 (CCL24)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Eotaxin-2 (CCL24) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Eotaxin-2 (CCL24) (500-P103G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Eotaxin-2 (CCL24). Western Blot: To detect hEotaxin-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hEotaxin-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P103GBT-1MG will be provided as 2 x 500 μg
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
Specifications
Eotaxin 2 | |
Polyclonal | |
Biotin | |
CCL24 | |
CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6369 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
O00175 | |
CCL24 | |
E.coli-derived Recombinant Human Eotaxin-2 (CCL24). | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.