Learn More
Invitrogen™ Eotaxin 2 Polyclonal Antibody
Description
For neutralization, use at a concentration of 7-8 μg/mL to achieve half of the neutralizing dose (ND50) of a 100.0 ng/mL concentration.
Specifications
Specifications
| Antigen | Eotaxin 2 |
| Applications | ELISA, Functional Assay, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CCL24 |
| Gene Accession No. | Q9JKC0 |
| Gene Alias | CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 |
| Gene Symbols | CCL24 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.