Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Eotaxin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104146
Description
Antibody detects endogenous levels of total CCL26.
Eotaxin-3 is a small cytokine belonging to the CC chemokine family that is also called CCL26, Chemokine (C-C motif) ligand 26. Chemokines play pivotal roles in health and disease by controlling migration and tissue localization of specific types of cells expressing their cognate receptors. Eotaxin-3 is chemotactic for eosinophils and basophils and elicits its effects by binding to the cell surface chemokine receptor CCR3. It is expressed by several tissues including heart, lung and ovary, and in endothelial cells that have been stimulated with the cytokine interleukin 4. This gene for chemokine is located on human chromosome 7. Eotaxin-3 induced migration of eosinophils and basophils only at a 10-fold higher concentration than eotaxin.
Specifications
Eotaxin 3 | |
Polyclonal | |
Unconjugated | |
CCL26 | |
CC chemokine IMAC; C-C motif chemokine 26; C-C motif chemokine ligand 26; Ccl26; Ccl26l; chemokine (C-C motif) ligand 26; chemokine N1; Eotaxin3; eotaxin-3; IMAC; Macrophage inflammatory protein 4-alpha; MGC126714; MIP-4a; MIP-4alpha; MIP-4-alpha; PRO242; SCYA26; SCYA26MIP-4-alpha; small inducible cytokine A26; small inducible cytokine subfamily A (Cys-Cys), member 26; small-inducible cytokine A26; Thymic stroma chemokine-1; TSC-1; UNQ216; UNQ216/PRO242 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10344 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y258 | |
CCL26 | |
A synthesized peptide derived from human CCL26(Accession Q9Y258), corresponding to amino acid residues L4-S54. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction