Learn More
Invitrogen™ Eotaxin Monoclonal Antibody, PeproTech®
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 500M25500UG

Description
AA Sequence of recombinant protein: GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP. Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human Eotaxin (CCL11). Anti-Human Eotaxin (CCL11)-specific antibody was purified from cell culture by Protein A affinity chromatography.
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
Eotaxin | |
Monoclonal | |
PBS with no preservative | |
P51671 | |
CCL11 | |
E.coli-derived Recombinant Human Eotaxin (CCL11). | |
500 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
Unconjugated | |
CCL11 | |
C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11 | |
Mouse | |
Protein A | |
RUO | |
6356 | |
-20°C | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.