Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eotaxin Mouse anti-Human, Clone: 13H2, MilliporeSigma™

Catalog No. MABC170425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC170425 25 μg
MABC1704100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC170425 Supplier MilliporeSigma Supplier No. MABC170425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Eotaxin Monoclonal antibody specifically detects Eotaxin in Human samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eotaxin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 13H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.
Host Species Mouse
Immunogen Full-length recombinant mature human Eotaxin
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.