Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Eotaxin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519868
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519868 Supplier Invitrogen™ Supplier No. PA519868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western blot applications, a monomer and homodimer are detected at ~9 and ~18kDa. A suggested positive control is Hela cell lysate. PA5-19868 can be used with blocking peptide PEP-0006.

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eotaxin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CCL11
Gene Accession No. P51671
Gene Alias C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Gene Symbols CCL11
Host Species Rabbit
Immunogen A peptide corresponding to amino acids near the carboxy terminus of human eotaxin.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6356
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.