Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ 384 Racks - Filter Pipette Tips
GREENER_CHOICE

Catalog No. 05414042 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.1 to 20 μL
5 to 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-414-042 5 to 100 μL
05-414-041 0.1 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-414-042 Supplier Eppendorf™ Supplier No. 0030078861
Add to Cart
Add to Cart

Eppendorf ep Dualfilter T.I.P.S.™ 384 Racks deliver cutting-edge filter pipette tips in 'PCR clean and Sterile' grade, specifically designed for 384-well plate applications, ensuring utmost precision and contamination protection in high-throughput labs.

  • ep Dualfilter T.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16- and 24-channel pipettes as well as 20 μL and 100 μL Move It adjustable tip spacing pipettes for utmost safety and reliability
  • Innovative SOFTattach technology allows for reliable tip fit and tightness while operating forces are reduced to a minimum
  • Fill a whole 384-well plate without loosening pipette tips
  • A perfect tip alignment is guaranteed by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of ep Dualfilter T.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Dual protection for pipette and sample against aerosols (1) and biomolecules
  • Lot-specific certificates and certificates of filter efficiency acc. to EN 29463 are available
  • Available as racks and in purity grades PCR clean and sterile
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 20 % lower polypropylene consumption per rack
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
  • (1) An aerosol is a colloid of fine solid particles or liquid droplets in air or another gas.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 5 to 100 μL
Volume (Metric) Dispensing 5 to 100 μL
Filtered Filtered
Sterility Sterile
Color Colorless
Rack Color Light Yellow
Autoclavable Not Autoclavable
Length (English) 2.1 in.
Length (Metric) 53 mm
Certifications/Compliance EN ISO 8655
Clean Claims PCR Clean and Sterile
For Use With (Application) DNA, RNA, Protein, Cell Culture Applications
Features PCR Clean
Format Rack
Tip Style Fine Point
Packaging 10 Racks x 384 Tips
Quantity 3840 Tips
Product Type Filter Pipette Tips
Includes 10 Racks x 384 Tips
Green Features Rack Bodies Made from Single-source Intra-type Collection PP Recyclate, Packaging Material Consists of at Least 37% Recycled Cardboard for Shipping Carton and at Least 60% for Folding Carton.
Color Code Light Yellow
ACT Label No
Disposable Single-use
For Use With (Equipment) Air-Cushion System
Product Line ep DualFilter T.I.P.S
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.