Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ 384, PCR Clean and Sterile

Catalog No. 05413614 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
100 to 0.5 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-413-614 100 to 0.5 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-413-614 Supplier Eppendorf™ Supplier No. 0030078411

ep Dualfilter T.I.P.S. 384 with the innovative SOFTattach technology are optimally coordinated to the Eppendorf 16 and 24-channel pipettes Research plus and Xplorer plus. Process 384-well plates manually with highest level of filter pipette tip tightness and coaxiality but extraordinary low operating forces.

  • ep Dualfilter T.I.P.S. 384 pipette tips build a system together with Research plus and Xplorer plus 16 and 24-channel pipettes for utmost safety and reliability
  • Innovative SOFTattach technology allows for reliable tip fit and tightness while operating forces are reduced to a minimum
  • Fill a whole 384-well plate without loosening pipette tips
  • A perfect tip alignment is guaranteed by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of ep Dualfilter T.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Dual protection for pipette and sample against aerosols and biomolecules
  • Lot-specific certificates are available at: (URL: https://www.eppendorf.com/certificates)[certificates homepage].
  • Certificate of filter efficiency acc. to EN 1822 available at: (URL: https://www.eppendorf.com/certificates)[certificates homepage]
  • Available as racks
  • Available in purity grades of: PCR clean and sterile
  • 1) An aerosol is a colloid of fine solid particles or liquid droplets in air or another gas.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 0.5 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model ep Dualfilter T.I.P.S.™ 384
Length (Metric) 53 mm
Clean Claims PCR clean and sterile, Forensic DNA Grade, Eppendorf Quality™
Storage Requirements Room temperature
Format Rack
Tip Style Universal
Packaging 10 racks × 384 tips
Quantity 3,840 tips
Color Code Light Yellow
Disposable Single-use
Product Line ep DualFilter T.I.P.S
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.