Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPA Phthalate Esters Mix, Certified Reference Material, 2,000 μg/mL Each Component in Hexane, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 mL
Specifications
Specifications
Packaging | Ampule |
Product Line | TraceCERT |
Concentration | 2,000 μg/mL each component in hexane |
Type | Organic Pollutant Standard |
Quantity | 1 mL |
Grade | TraceCERT™ Analytical Standard |
Concentration or Composition (by Analyte or Components) | Bis(2-ethylhexyl) phthalate; Benzyl butyl phthalate; Dibutyl phthalate; Di-n-octyl phthalate; Diethyl phthalate; Dimethyl phthalate |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction