Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epac2 Antibody (SR1071), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | Epac2 |
---|---|
Clone | SR1071 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Epac2 Monoclonal antibody specifically detects Epac2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Epac2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CAMP-GEFII, cAMP-regulated guanine nucleotide exchange factor II, CGEF2EPAC 2, EPAC2, Exchange factor directly activated by cAMP 2, Exchange protein directly activated by cAMP 2, Nbla00496, putative protein product of Nbla00496, Rap guanine nucleotide exchange factor (GEF) 4, rap guanine nucleotide exchange factor 4, RAP guanine-nucleotide-exchange factor (GEF) 4 | |
A synthesized peptide derived from human Epac2 (Uniprot #: Q8WZA2) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1071 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
11069 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title