Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPAC2 Rabbit anti-Equine, Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206903
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6903 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6903 Supplier Fabgennix Inc Supplier No. EPAC201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPAC2 Polyclonal antibody specifically detects EPAC2 in Equine, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen EPAC2
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Rapgef4
Gene Accession No. Q8WZA2, Q9EQZ6, Q9Z1C7
Gene Alias 1300003D15Rik; 5730402K07Rik; 6330581N18Rik; cAMP-dependent Rap1 guanine-nucleotide exchange factor; cAMP-GEFII; cAMP-regulated guanine nucleotide exchange factor II; cAMP-regulated guanine nucleotide exchange factor IIB; CAMPS; Cgef2; EPAC; epac 2; Epac2; Epac-2; Exchange factor directly activated by cAMP 2; exchange protein directly activated by cAMP 2; gefii; mKIAA4040; Nbla00496; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; Rap guanine nucleotide exchange factor 4; RAP guanine-nucleotide-exchange factor (GEF) 4; Rapgef4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 743-762 of Epac2 protein.Sequence:CPREQFDSLTPLPEQEGPT
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100064121, 11069, 252857, 56508
Target Species Equine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.