Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPAS1 Mouse anti-Human, Clone: 2F6B12, Proteintech
SDP

Catalog No. 501737143 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7143 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7143 Supplier Proteintech Group Inc Supplier No. 667311IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.

Specifications

Antigen EPAS1
Applications Western Blot
Classification Monoclonal
Clone 2F6B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EPAS1
Gene Accession No. Q99814
Gene Alias bHLHe73, ECYT4, EPAS 1, EPAS1, HIF 1 alpha like factor, HIF 2 alpha, HIF2 alpha, HIF2A, HLF, Member of PAS protein 2, MOP2, PASD2
Gene Symbols EPAS1
Host Species Mouse
Immunogen EPAS1 Fusion Protein Ag24886
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2034
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.