Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565015 Supplier Invitrogen™ Supplier No. PA565015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESVPEPRPSEW DKRLSTHSPF RTLNINGQIP TGEGPPLVKT QTVTISDNAN AVKSEIPTKD VPIVHTETKT Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal dominant, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4. 1. 'Rh-unlinked' forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPB41
Gene Accession No. P11171
Gene Alias 4.1R; AI415518; Band 4.1; D4Ertd442e; E41P; EL1; elliptocytosis 1, RH-linked; Elp1; Elp-1; EPB4.1; EPB41; erythrocyte membrane protein band 4.1; erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked); erythrocyte protein band 4.1; erythrocyte skeletal protein; erythrocyte surface protein band 4.1; HE; Kiaa4056; mKIAA4056; P4.1; protein 4.1; Protein 4.1R; RGD1564762
Gene Symbols EPB41
Host Species Rabbit
Immunogen Recombinant Human EPB41. Recombinant protein control fragment (Product #RP-104584).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.