Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPB41 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588540 Supplier Thermo Scientific Supplier No. PA588540

Rabbit Polyclonal Antibody

Immunogen sequence: MHCKVSLLDD TVYECVVEKH AKGQDLLKRV CEHLNLLEED YFGLAIWDNA TSKTWLDSAK EIKKQVRGVP WNFTFNVKFY PPDPAQLTED ITRYYLCLQL RQDIVAGRLP CSFATLALLG SYTIQSELGD YDPELHGVDY VSDFKLAPNQ TKELEEKVME LHKSYRSMTP AQADLEFLEN AKKLSMYGVD LHKAKDLEGV DIILGVCSSG LLVYKDKLRI NRFPWPKVLK ISYKRSSFFI KIRPGEQEQY ESTIGFKLPS YRAAKKLWKV CVEHHTFFRL TSTDTIPKSK F; Positive Samples: Mouse brain, Mouse liver; Cellular Location: Cytoplasm, Nucleus, cell cortex, cytoskeleton

Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal dominant, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4. 1. 'Rh-unlinked' forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41
Applications Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene EPB41
Gene Accession No. P11171, P48193
Gene Alias 4.1R, AI415518, Band 4.1, D4Ertd442e, E41P, EL1, elliptocytosis 1, RH-linked, Elp1, Elp-1, EPB4.1, EPB41, erythrocyte membrane protein band 4.1, erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked), erythrocyte protein band 4.1, erythrocyte skeletal protein, erythrocyte surface protein band 4.1, HE, Kiaa4056, mKIAA4056, P4.1, protein 4.1, Protein 4.1R, RGD1564762
Gene Symbols EPB41
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-500 of human EPB41 (NP_0011594781)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2035, 269587, 313052
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.