Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPB41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 130141AP150UL

Description
Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal domit, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4.1. ′Rh-unlinked′ forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.Specifications
EPB41 | |
Polyclonal | |
Unconjugated | |
EPB41 | |
4.1R, Band 4.1, E41P, EL1, EPB4.1, EPB41, HE, P4.1, Protein 4.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2035, 269587, 313052 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11171, P48193 | |
EPB41 | |
EPB41 Fusion Protein Ag4075 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction