Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562918 Supplier Invitrogen™ Supplier No. PA562918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAEVDFTVIG DYHGSAFEDF SRSLPELDRD KSDSDTEGLL FSRDLNKGAP SQDDESGGIE DSPDRGACST PDMPQFEPVK TETMTVSSLA IRKKIEPE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPB41L1
Gene Accession No. Q9H4G0
Gene Alias 4.1N; band 4.1-like protein 1; DKFZp686H17242; Epb4; Epb4.1l1; Epb41l1; erythrocyte membrane protein band 4.1 like 1; erythrocyte membrane protein band 4.1-like 1; erythrocyte protein band 4.1 like 1; erythrocyte protein band 4.1-like 1; KIAA0338; MGC11072; mKIAA0338; MRD11; NBL1; neuronal protein 4.1; neuron-type nonerythroid protein 4.1; RP11-234K24.1
Gene Symbols EPB41L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPB41L1. Recombinant protein control fragment (Product #RP-99295).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.