Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPB41L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563352
Description
Immunogen sequence: KKAQEEAPQQ PEAAAAVTTP VTPAGHGHPE ANSNEKHPSQ QDTRPAEQSL DMEEKDYSEA DGLSERTTPS KAQKSPQKIA K Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane.
Specifications
EPB41L1 | |
Polyclonal | |
Unconjugated | |
EPB41L1 | |
4.1N; band 4.1-like protein 1; DKFZp686H17242; Epb4; Epb4.1l1; Epb41l1; erythrocyte membrane protein band 4.1 like 1; erythrocyte membrane protein band 4.1-like 1; erythrocyte protein band 4.1 like 1; erythrocyte protein band 4.1-like 1; KIAA0338; MGC11072; mKIAA0338; MRD11; NBL1; neuronal protein 4.1; neuron-type nonerythroid protein 4.1; RP11-234K24.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4G0 | |
EPB41L1 | |
Recombinant protein corresponding to Human EPB41L1. Recombinant protein control fragment (Product #RP-99294). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction