Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579206
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579206 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579206 Supplier Invitrogen™ Supplier No. PA579206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human RT4 whole cell, human Caco-2 whole cell.

Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene EPB41L1
Gene Accession No. Q9H4G0, Q9WTP0, Q9Z2H5
Gene Alias 4.1N; band 4.1-like protein 1; DKFZp686H17242; Epb4; Epb4.1l1; Epb41l1; erythrocyte membrane protein band 4.1 like 1; erythrocyte membrane protein band 4.1-like 1; erythrocyte protein band 4.1 like 1; erythrocyte protein band 4.1-like 1; KIAA0338; MGC11072; mKIAA0338; MRD11; NBL1; neuronal protein 4.1; neuron-type nonerythroid protein 4.1; RP11-234K24.1
Gene Symbols EPB41L1
Host Species Rabbit
Immunogen E.coli-derived human EPB41L1 recombinant protein (Position: Q691-S881).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13821, 2036, 59317
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.