Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPB41L2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518821
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
The 4.1 gene family encodes a group of multifunctional cytoskeletal proteins (4.1R, 4.1G, 4.1N and 4.1B), which are predominantly expressed in the nervous system. 4.1G is a protein that stabilizes spectrin-actin interactions and is associated with hereditary elliptocytosis. Red blood cell 4.1, designated 4.1R, is a multifunctional protein that is essential for maintaining erythrocyte shape and membrane mechanical properties. Both 4.1R and 4.1G are distributed in a unique pattern in the cerebellum and are believed to modulate the membrane mechanical properties of neuronal cells by promoting fodrin/actin association. 4.1N and 4.1B, designated EPB41L1 and EPB41L3, respectively, are strongly expressed in the brain. Antibodies to 4.1N have been reported to detect multiple forms, each enriched in postsynaptic density preparations relative to brain homogenate. Antibodies to 4.1B have been reported to detect two forms.
Specifications
EPB41L2 | |
Polyclonal | |
Unconjugated | |
EPB41L2 | |
4.1G; 4.1-G; AW555191; band 4.1-like protein 2; D10Ertd398e; DKFZp781D1972; DKFZp781H1755; Epb4.1l2; Epb41l2; erythrocyte membrane protein band 4.1 like 2; erythrocyte membrane protein band 4.1 like-protein 2; erythrocyte membrane protein band 4.1-like 2; erythrocyte membrane protein band 4.1-like 2 retinal; erythrocyte protein band 4.1 like 2; erythrocyte protein band 4.1l2 long; erythrocyte protein band 4.1l2 middle; erythrocyte protein band 4.1-like 2; generally expressed protein 4.1; NBL2; OTTHUMP00000040262; RGD1563977; RP3-324N14.1 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
2037 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43491 | |
EPB41L2 | |
Synthetic peptide sequence (RREVRSPTKAPH) corresponding to the internal amino acids of EPB41L2 (aa 593-605). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction