Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529485 Supplier Invitrogen™ Supplier No. PA529485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4. Predicted reactivity: Mouse (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The 4.1 gene family encodes a group of multifunctional cytoskeletal proteins (4.1R, 4.1G, 4.1N and 4.1B), which are predominantly expressed in the nervous system. 4.1G is a protein that stabilizes spectrin-actin interactions and is associated with hereditary elliptocytosis. Red blood cell 4.1, designated 4.1R, is a multifunctional protein that is essential for maintaining erythrocyte shape and membrane mechanical properties. Both 4.1R and 4.1G are distributed in a unique pattern in the cerebellum and are believed to modulate the membrane mechanical properties of neuronal cells by promoting fodrin/actin association. 4.1N and 4.1B, designated EPB41L1 and EPB41L3, respectively, are strongly expressed in the brain. Antibodies to 4.1N have been reported to detect multiple forms, each enriched in postsynaptic density preparations relative to brain homogenate. Antibodies to 4.1B have been reported to detect two forms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L2
Applications Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene EPB41L2
Gene Accession No. O43491
Gene Alias 4.1G; 4.1-G; AW555191; band 4.1-like protein 2; D10Ertd398e; DKFZp781D1972; DKFZp781H1755; Epb4.1l2; Epb41l2; erythrocyte membrane protein band 4.1 like 2; erythrocyte membrane protein band 4.1 like-protein 2; erythrocyte membrane protein band 4.1-like 2; erythrocyte membrane protein band 4.1-like 2 retinal; erythrocyte protein band 4.1 like 2; erythrocyte protein band 4.1l2 long; erythrocyte protein band 4.1l2 middle; erythrocyte protein band 4.1-like 2; generally expressed protein 4.1; NBL2; OTTHUMP00000040262; RGD1563977; RP3-324N14.1
Gene Symbols EPB41L2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EPB41L2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.