Learn More
EPB41L3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NB11061027AF350
Catalog No. N1061027A30
Only null left
Add to Cart
Description
Description
EPB41L3 Polyclonal specifically detects EPB41L3 in Human, Mouse, Rat, Primate, Xenopus samples. It is validated for Western Blot.
Specifications
Specifications
EPB41L3
Polyclonal
Western Blot
4.1BFLJ37633, DAL1DAL-1,, Differentially expressed in adenocarcinoma of the lung protein 1, erythrocyte membrane protein band 4.1-like 3, KIAA0987band 4.1-like protein 3
Rabbit
0.1 mL
23136
Store at 4°C in the dark.
Western Blot
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
EPB41L3
A synthetic peptide made to an internal region of human EPB41L3 (within residues 400-500). [Swiss-ProtNo. Q9Y2J2]
Primary
Human, Mouse, Rat, Primate, Xenopus species
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.