Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPB41L3 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061027AF594
Description
EPB41L3 Polyclonal specifically detects EPB41L3 in Human, Mouse, Rat, Primate, Xenopus samples. It is validated for Western Blot.Specifications
EPB41L3 | |
Polyclonal | |
Western Blot | |
4.1BFLJ37633, DAL1DAL-1,, Differentially expressed in adenocarcinoma of the lung protein 1, erythrocyte membrane protein band 4.1-like 3, KIAA0987band 4.1-like protein 3 | |
Rabbit | |
0.1 mL | |
23136 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
EPB41L3 | |
A synthetic peptide made to an internal region of human EPB41L3 (within residues 400-500). [Swiss-ProtNo. Q9Y2J2] | |
Primary | |
Human, Mouse, Rat, Primate, Xenopus species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction