Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPB41L3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061027PCP
Description
EPB41L3 Polyclonal antibody specifically detects EPB41L3 in Human, Mouse, Rat, Primate, Xenopus samples. It is validated for Western Blot, Western BlotSpecifications
EPB41L3 | |
Polyclonal | |
Western Blot, Simple Western | |
4.1BFLJ37633, DAL1DAL-1,, Differentially expressed in adenocarcinoma of the lung protein 1, erythrocyte membrane protein band 4.1-like 3, KIAA0987band 4.1-like protein 3 | |
A synthetic peptide made to an internal region of human EPB41L3 (within residues 400-500). [Swiss-Prot# Q9Y2J2] | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Signal Transduction, Tumor Suppressors | |
23136 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction