Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA146313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA146313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA146313 Supplier Invitrogen™ Supplier No. PA146313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This EPB41L3 antibody is useful for Western blot, where bands are seen at ~120 and 96 kDa. It detects both isoforms A and B in mouse brain lysates. It detects human isoform A strongly and isoform B more weakly.Suggested positive control: human and mouse brain lysate, HeLa whole cell lysate, Hela whole cell lysate, mouse brain lysate, human brain lysate.

Critical growth regulator in the pathogenesis of meningiomas. Its human homolog acts as a tumor suppressor and binds 14-3-3 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L3
Applications Western Blot
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.1% sodium azide
Gene EPB41L3
Gene Accession No. Q9WV92, Q9Y2J2
Gene Alias 130kDa Protein 4.1B MEF cell isoform; 4.1B; 60kDa 4.1B MEF cell isoform; band 4.1-like protein 3; Band 4.1-like protein 3, N-terminally processed; Dal1; DAL-1; DAL1P; differentially expressed in adenocarcinoma of the lung protein 1; Epb4.1l3; Epb41l3; epb41l3.L; erythrocyte membrane protein band 4.1 like 3; erythrocyte membrane protein band 4.1-like 3; erythrocyte membrane protein band 4.1-like 3 L homeolog; erythrocyte protein band 4.1 like 3; erythrocyte protein band 4.1-like 3; FLJ37633; HGNC:3380; Kiaa0987; mDAL-1; NBL3; XELAEV_18031863mg
Gene Symbols EPB41L3, epb41l3.L
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human EPB41L3 (within residues 400-500).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116724, 13823, 23136, 398970
Target Species Human, Mouse, Rat, Monkey, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.