Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527528

Catalog No. PIPA527528


Only null left
Add to Cart

Description

Description

Recommended positive controls: PC-3, U87-MG. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (86%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Critical growth regulator in the pathogenesis of meningiomas. Its human homolog acts as a tumor suppressor and binds 14-3-3 proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EPB41L3
Polyclonal
Unconjugated
EPB41L3
130kDa Protein 4.1B MEF cell isoform; 4.1B; 60kDa 4.1B MEF cell isoform; band 4.1-like protein 3; Band 4.1-like protein 3, N-terminally processed; Dal1; DAL-1; DAL1P; differentially expressed in adenocarcinoma of the lung protein 1; Epb4.1l3; Epb41l3; epb41l3.L; erythrocyte membrane protein band 4.1 like 3; erythrocyte membrane protein band 4.1-like 3; erythrocyte membrane protein band 4.1-like 3 L homeolog; erythrocyte protein band 4.1 like 3; erythrocyte protein band 4.1-like 3; FLJ37633; HGNC:3380; Kiaa0987; mDAL-1; NBL3; XELAEV_18031863mg
Rabbit
Antigen affinity chromatography
RUO
23136
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.97 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q9Y2J2
EPB41L3
Recombinant protein encompassing a sequence within the C-terminus region of human EPB41L3. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.