Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583023 Supplier Invitrogen™ Supplier No. PA583023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TESSGIETEP TVHHLPLSTE KVVQETVLVE ERRVVHASGD ASYSAGDSGD AAAQPAFTGI KGKEGSALTE GAKEEGGEEV AKAVLE Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 58%.

Critical growth regulator in the pathogenesis of meningiomas. Its human homolog acts as a tumor suppressor and binds 14-3-3 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPB41L3
Gene Accession No. Q9Y2J2
Gene Alias 130kDa Protein 4.1B MEF cell isoform; 4.1B; 60kDa 4.1B MEF cell isoform; band 4.1-like protein 3; Band 4.1-like protein 3, N-terminally processed; Dal1; DAL-1; DAL1P; differentially expressed in adenocarcinoma of the lung protein 1; Epb4.1l3; Epb41l3; epb41l3.L; erythrocyte membrane protein band 4.1 like 3; erythrocyte membrane protein band 4.1-like 3; erythrocyte membrane protein band 4.1-like 3 L homeolog; erythrocyte protein band 4.1 like 3; erythrocyte protein band 4.1-like 3; FLJ37633; HGNC:3380; Kiaa0987; mDAL-1; NBL3; XELAEV_18031863mg
Gene Symbols EPB41L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPB41L3. Recombinant protein control fragment (Product #RP-93121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.