Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPB41L3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | EPB41L3 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6537
![]() ![]() |
Proteintech Group Inc
107191AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Protein 4. 1B is a member of a family of similar proteins whose expression is frequently lost in a variety of human tumors, including meningiomas, non-small-cell lung cancers, and breast carcinomas and is frequently down-regulated in human clinical prostate cancer. Studies have shown that Protein 4. 1B normally triggers death in metastatic cells, thereby preventing cancer from spreading. A clinical test for Protein 4. 1B activity could be useful in assessing the metastatic potential of prostate cancers.Specifications
EPB41L3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9WV92, Q9Y2J2 | |
116724, 13823, 23136 | |
EPB41L3 Fusion Protein Ag1089 | |
Primary | |
-20°C | |
EPB41L3 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
4.1B, Band 4.1 like protein 3, DAL 1, DAL1, EPB41L3, KIAA0987 | |
EPB41L3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title