Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557728 Supplier Invitrogen™ Supplier No. PA557728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNSLSRKLSK FGSIRYKHRY SGRTALQMSR DLSIQLPRPD QNVTRSRSKT YPKRIAQTQP AESNSISRIT ANMENGENEG TIK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L4A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPB41L4A
Gene Accession No. Q9HCS5
Gene Alias Band 4.1-like protein 4A; Epb4.1l4; Epb4.1l4a; EPB41L4; Epb41l4a; erythrocyte membrane protein band 4.1 like 4A; erythrocyte protein band 4.1 like 4a; erythrocyte protein band 4.1-like 4; erythrocyte protein band 4.1-like 4a; NBL4; Protein NBL4
Gene Symbols EPB41L4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPB41L4A. Recombinant protein control fragment (Product #RP-96842).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.