Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPB41L4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549596
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549596 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549596 Supplier Thermo Scientific Supplier No. PA549596
Add to Cart
Add to Cart

EPB41L4B Polyclonal Antibody

EPB41L4B Polyclonal Antibody for Western Blot, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L4B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EPB41L4B
Gene Accession No. Q9H329
Gene Alias 6430543G08Rik; AA589614; AU021054; band 4.1-like protein 4B; BB007528; CG1; D4Ertd346e; Ehm2; Epb4.1l4b; EPB41L4B; erythrocyte membrane protein band 4.1 like 4B; erythrocyte protein band 4.1 like 4b; erythrocyte protein band 4.1-like 4b; erythrocyte protein band 4.1-like 4b; expressed in high-metastatic cells; expressed in high-metastatic cells; FERM-containing protein CG1; LULU2; Protein EHM2; RGD1562988
Gene Symbols EPB41L4B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 650-677 amino acids from the C-terminal region of human EPB41L4B.
Purification Method Ammonium Sulfate Precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54566
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.