Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPB41L4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560150
Description
Immunogen sequence: PLHININKAE EKKVSEKTLQ TPLLPSPVAD HVKCNILKAQ LENASRVNIQ GGKEESPFVN INKKSSLQDA SVRSPIPIRV ETAQPAVEKP EIKPPRVRKL TRQYSFDEDD LPP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Up-regulates the activity of the Rho guanine nucleotide exchange factor ARHGEF18. Involved in the regulation of the circumferential actomyosin belt in epithelial cells (PubMed:22006950). Promotes cellular adhesion, migration and motility in vitro and may play a role in wound healing (PubMed:23664528). May have a role in mediating cytoskeletal changes associated with steroid-induced cell differentiation (PubMed:14521927). [UniProt].
Specifications
EPB41L4B | |
Polyclonal | |
Unconjugated | |
EPB41L4B | |
6430543G08Rik; AA589614; AU021054; band 4.1-like protein 4B; BB007528; CG1; D4Ertd346e; Ehm2; Epb4.1l4b; EPB41L4B; erythrocyte membrane protein band 4.1 like 4B; erythrocyte protein band 4.1 like 4b; erythrocyte protein band 4.1-like 4b; erythrocyte protein band 4.1-like 4b; expressed in high-metastatic cells; expressed in high-metastatic cells; FERM-containing protein CG1; LULU2; Protein EHM2; RGD1562988 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H329 | |
EPB41L4B | |
Recombinant protein corresponding to Human EPB41L4B. Recombinant protein control fragment (Product #RP-92888). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction