Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB41L4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142309
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142309 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142309 Supplier Invitrogen™ Supplier No. PA5142309
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise both reported isoforms (NP_060894.2; NP_061987.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

Up-regulates the activity of the Rho guanine nucleotide exchange factor ARHGEF18. Involved in the regulation of the circumferential actomyosin belt in epithelial cells (PubMed:22006950). Promotes cellular adhesion, migration and motility in vitro and may play a role in wound healing (PubMed:23664528). May have a role in mediating cytoskeletal changes associated with steroid-induced cell differentiation (PubMed:14521927). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene EPB41L4B
Gene Accession No. Q9H329
Gene Alias 6430543G08Rik; AA589614; AU021054; band 4.1-like protein 4B; BB007528; CG1; D4Ertd346e; Ehm2; Epb4.1l4b; EPB41L4B; erythrocyte membrane protein band 4.1 like 4B; erythrocyte protein band 4.1 like 4b; erythrocyte protein band 4.1-like 4b; erythrocyte protein band 4.1-like 4b; expressed in high-metastatic cells; expressed in high-metastatic cells; FERM-containing protein CG1; LULU2; Protein EHM2; RGD1562988
Gene Symbols EPB41L4B
Host Species Goat
Immunogen Peptide with sequence C-HNYQPQYHPNIHPSQ.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54566
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.