Learn More
Invitrogen™ EPB41L5 Polyclonal Antibody

Description
Predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | EPB41L5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EPB41L5 |
| Gene Accession No. | Q9HCM4 |
| Gene Alias | Band 4.1-like protein 5; BE37; EPB41L5; erythrocyte membrane protein band 4.1 like 5; KIAA1548; YMO1; YRT; yurt homolog |
| Gene Symbols | EPB41L5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.