Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPB42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559127 Supplier Invitrogen™ Supplier No. PA559127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAQEDIAICR PHLAIKMPEK AEQYQPLTAS VSLQNSLDAP MEDCVISILG RGLIHRERSY RFRSVWPENT MCAKFQFTPT HVGLQRLTVE VDCNMFQNLT NYKSVT Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 68%.

Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB42
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPB42
Gene Accession No. P16452
Gene Alias BEP42; E42P; Epb4.2; Epb42; Erythrocyte membrane protein band 4.2; erythrocyte protein 4.2; erythrocyte protein band 4.2; erythrocyte surface protein band 4.2; membrane skeletal protein; P4.2; PA; SPH5
Gene Symbols EPB42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPB42. Recombinant protein control fragment (Product #RP-98173).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.