Learn More
Invitrogen™ EPB42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109686
Description
Immunogen sequence: CDFQAARNNE EHHTKALSSR RLFVRRGQPF TIILYFRAPV RAFLPALKKV ALTAQTGEQP SKINRTQATF PISSLGDRKW WSAVVEERDA QSWTISVTTP ADAVIGHYSL LLQVSGRKQL LLGQFTLLFN PWNREDAVFL KNEAQRMEYL LNQNGLIYLG TADCIQAESW DFGQFEGDVI DLSLRLLSKD KQVEKWSQPV HVARVLGALL HFLKEQRVLP TPQTQATQEG A.
Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| EPB42 | |
| Polyclonal | |
| Unconjugated | |
| EPB42 | |
| BEP42; E42P; Epb4.2; Epb42; Erythrocyte membrane protein band 4.2; erythrocyte protein 4.2; erythrocyte protein band 4.2; erythrocyte surface protein band 4.2; membrane skeletal protein; P4.2; PA; SPH5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13828, 2038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P16452, P49222 | |
| EPB42 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-240 of human EPB42 (NP_001107606). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.