Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145011
Description
Immunogen sequence: SALLQPSNIT QTSSSHSALS HQVTAANSAT TQVLIGNNIR LTVPSSVATV NSIAPINARH IPRTLSAVPS SALKLAAAAN CQ Antigen sequence identity: Mouse 89%; Rat 94%.
EPC1 is a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. NuA4 may also play a direct role in DNA repair when directly recruited to sites of DNA damage.
Specifications
EPC1 | |
Polyclonal | |
Unconjugated | |
EPC1 | |
2400007E14Rik; 5730566F07Rik; A930032N02Rik; DKFZp781P2312; enhancer of polycomb 1; enhancer of polycomb homolog 1; enhancer of polycomb homolog 1 (Drosophila); Epc1; Epl 1; Epl1; mir-1893 | |
Rabbit | |
Affinity chromatography | |
RUO | |
80314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2F5 | |
EPC1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 717-798. Recombinant protein control fragment (Product #RP-109888). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction