Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557802
Description
Immunogen sequence: YATPATLHNG NHHKVQECKT KHPHHLSLKE EASDVVRQKK KYPKKPKAEA LITSQQPTPE TLPVINKSDI KQYDFHSSDE DEFPQVLS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 89%.
EPC2 may play a role in transcription or DNA repair.
Specifications
EPC2 | |
Polyclonal | |
Unconjugated | |
EPC2 | |
5830499L14; D2Ertd694e; Enhancer of polycomb homolog 2; enhancer of polycomb homolog 2 (Drosophila); Epc2; EPC-LIKE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q52LR7 | |
EPC2 | |
Recombinant protein corresponding to Human EPC2. Recombinant protein control fragment (Product #RP-96086). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction