Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpCAM (CD326) Monoclonal Antibody (G8.8), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544217
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4217 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4217 Supplier Leinco Technologies Inc Supplier No. C724100MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: Mouse EpCAM (CD326). Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month.

EpCAM (CD326) Monoclonal antibody specifically detects EpCAM (CD326) in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen EpCAM (CD326)
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G8.8
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene EPCAM
Gene Accession No. P16422, Q99JW5
Gene Alias adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1
Host Species Rat
Immunogen TE-71 thymic epithelial cell line
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17075, 4072
Target Species Human, Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.