Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPCAM Mouse anti-Human, Clone: EGP40/1373, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14586
Description
Mouse monoclonal antibody raised against partial recombinant human EPCAM.
- High specificity
- High reactivity
Specifications
EPCAM | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
P16422 | |
EPCAM | |
Recombinant protein corresponding to amino acids 77-202 at extracellular domain of human EPCAM. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
EGP40/1373 | |
Mouse monoclonal antibody raised against partial recombinant human EPCAM. | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2 | |
Mouse | |
Protein A/G purification | |
RUO | |
4072 | |
Store at 4°C. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction